Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water

Catalog No. AAH2850003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2850003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2850003 Supplier Thermo Scientific Chemicals Supplier No. H2850003

missing translation for 'validMainframeMsg'

CAS: 101555-62-8 | C20H19NO4 | 337.375 g/mol

In organic synthesis, N-Fmoc-D-proline are often employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In organic synthesis, N-Fmoc-D-proline are often employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101555-62-8
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.375
MDL Number MFCD00077067
InChI Key ZPGDWQNBZYOZTI-GOSISDBHSA-N
Synonym fmoc-d-pro-oh, fmoc-d-proline, n-9h-fluoren-9-ylmethoxy carbonyl-d-proline, n-fmoc-d-proline, fmoc-d-pro, 2r-1-9h-fluoren-9-ylmethoxycarbonyl pyrrolidine-2-carboxylic acid, 2r-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, fmoc-3-pro-oh, ksc908s9r, n-alpha-fmoc-d-proline
PubChem CID 688136
IUPAC Name (2R)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O

Specifications

Melting Point 110°C to 116°C
Quantity 1 g
Formula Weight 337.38
Percent Purity ≥98%
Assay may contain up to ≈5% water
Chemical Name or Material N-Fmoc-D-proline
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.