Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-D-tyrosine, 97%

Catalog No. AAH66740MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66740MD 250 mg
AAH6674003 1 g
AAH6674006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66740MD Supplier Thermo Scientific Chemicals Supplier No. H66740MD

CAS: 112883-29-1 | C24H21NO5 | 403.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112883-29-1
Molecular Formula C24H21NO5
Molecular Weight (g/mol) 403.43
MDL Number MFCD00171384
InChI Key SWZCTMTWRHEBIN-ANBDAQEENA-N
Synonym fmoc-d-tyrosine, fmoc-d-tyr-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-hydroxyphenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-hydroxyphenyl propanoic acid, n-fmoc-d-tyr oh, fmoc-d-phe 4-oh-oh, n-9h-fluorene-9-ylmethoxycarbonyl-d-tyrosine, n-alpha-9-fluorenylmethoxycarbonyl-d-tyrosine, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-hydroxy-phenyl-propionic acid
PubChem CID 6957988
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-hydroxyphenyl)propanoic acid
SMILES OC(=O)[C@@H](CC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 175°C to 180°C
Color White
Quantity 250 mg
Formula Weight 403.43
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Fmoc-D-tyrosine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.