Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-L-aspartic acid 4-benzyl ester, 97%

Catalog No. AAH6201106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6201106 5 g
AAH6201114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6201106 Supplier Thermo Scientific Chemicals Supplier No. H6201106
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 86060-84-6 | C26H23NO6 | 445.47 g/mol

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Solubility
Soluble in methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86060-84-6
Molecular Formula C26H23NO6
Molecular Weight (g/mol) 445.47
MDL Number MFCD00065630
InChI Key OQGAELAJEGGNKG-UHFFFAOYNA-N
Synonym fmoc-asp obzl-oh, fmoc-l-aspartic acid 4-benzyl ester, l-fmoc-aspartic acid beta-benzyl ester, fmoc-l-asp obzl-oh, 2s-4-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-l-aspartic acid beta-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy-4-oxobutanoic acid, n-fmoc-l-aspartic acid beta-benzyl ester, fmoc-l-asp obn, fmoc-l-asp bzl-oh
PubChem CID 13966931
SMILES OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 120°C to 130°C
Quantity 5 g
Beilstein 4609615
Solubility Information Soluble in methanol.
IUPAC Name 4-(benzyloxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid
Formula Weight 445.46
Percent Purity 97%
Chemical Name or Material N-Fmoc-L-aspartic acid 4-benzyl ester
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.