Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-L-beta-homoleucine, 95%

Catalog No. AAH5197603
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5197603 1 g
AAH51976MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5197603 Supplier Thermo Scientific Chemicals Supplier No. H5197603
Only null left
Add to Cart

CAS: 193887-44-4 | C22H25NO4 | 367.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193887-44-4
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD01863059
InChI Key YLVSABQQLLRFIJ-HNNXBMFYSA-N
Synonym fmoc-beta-holeu-oh, fmoc-l-beta-homoleucine, fmoc-l-, a-homo-leu-oh, fmoc-beta-homoleu-oh, s-3-fmoc-amino-5-methylhexanoic acid, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-5-methylhexanoic acid, fmoc-?-holeu-oh, ambotzfaa6690, 3s-3-9h-fluoren-9-ylmethoxycarbonylamino-5-methylhexanoic acid, fmoc-holeu-oh fmoc-homoleucine
PubChem CID 2761527
IUPAC Name (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methylhexanoic acid
SMILES CC(C)C[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 7723942
Formula Weight 367.44
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-beta-homoleucine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.