Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-L-glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals

Catalog No. AAH6655106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6655106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6655106 Supplier Thermo Scientific Chemicals Supplier No. H6655106
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 122350-52-1 | Molecular Formula: C27H25NO6 | Molecular Weight: 459.498 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122350-52-1
Molecular Formula C27H25NO6
Molecular Weight (g/mol) 459.498
MDL Number MFCD00080272
InChI Key FMWLYDDRYGOYMY-DEOSSOPVSA-N
Synonym fmoc-glu-obzl, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, 4s-5-benzyloxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, fmoc-l-glutamic acid alpha-benzyl ester, nalpha-fmoc-l-glutamic acid alpha-benzyl ester
PubChem CID 10049854
IUPAC Name (4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Color White
Quantity 5 g
Formula Weight 459.5
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-glutamic acid 1-benzyl ester
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.