Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-L-homoarginine, 96%, Thermo Scientific™

Catalog No. AAH6322803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6322803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6322803 Supplier Thermo Scientific Chemicals Supplier No. H6322803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 776277-76-0
Molecular Formula C22H26N4O4
Molecular Weight (g/mol) 410.474
MDL Number MFCD01861360
InChI Key QRELIJBJYZHTQU-IBGZPJMESA-N
Synonym fmoc-hoarg-oh, fmoc-homoarg-oh, fmoc-har-oh, fmoc-l-homoarginine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-guanidinohexanoic acid, fmoc-homoarg, 2s-6-carbamimidamido-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, n∼6∼-diaminomethylidene-n∼2∼-9h-fluoren-9-yl methoxy carbonyl-l-lysine, 2s-6-diaminomethylidene amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid
PubChem CID 40418724
IUPAC Name (2S)-6-(diaminomethylideneamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCN=C(N)N)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 410.47
Percent Purity 96%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-homoarginine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.