Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-L-methionine sulfone, 98%, Thermo Scientific™

Catalog No. AAH63033MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63033MD 250 mg
AAH6303303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63033MD Supplier Thermo Scientific Chemicals Supplier No. H63033MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163437-14-7
Molecular Formula C20H21NO6S
Molecular Weight (g/mol) 403.45
MDL Number MFCD00153361
InChI Key KJLKPACOHZKRFM-SFHVURJKSA-N
PubChem CID 7016362
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methanesulfonylbutanoic acid
SMILES CS(=O)(=O)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Color White
Synonym fmoc-met o2-oh, fmoc-l-methionine sulfone, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylsulfonyl butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methanesulfonylbutanoic acid, ambotzfaa1406, fmoc-met o 2-oh, pubchem22746, n-alpha-9-fluorenylmethyloxycarbonyl-l-methioninesulfon, s-2-9h-fluoren-9-yl methoxy carbonyl-amino-4-methylsulfonyl butanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino-4-methylsulfonyl butanoic acid
Quantity 250 mg
Formula Weight 403.45
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-methionine sulfone
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.