Learn More
Medchemexpress LLC N-Fmoc-leucine-d3 | 538372-74-6 | 99.3% | 356.43 | C21H20D3NO4 | 100 MG
SDP
Catalog No. 5000213103
Encompass_Preferred
Deuterated Fmoc-protected leucine is a stable-isotope labeled amino acid derivative used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is supplied as a white to off-white solid of high purity suitable for analytical and research applications.
  • Deuterium-labeled Fmoc-protected leucine for isotope-dilution quantitation.
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • High purity (≈99.3%) for accurate analytical results.
  • White to off-white solid form for easy handling and storage.
  • Molecular weight 356.43 and CAS number 538372-74-6.
Catalog No. 50-002-13103 Supplier Medchemexpress LLC Supplier No. HY101064S2100MG
Deuterated Fmoc-protected leucine is a stable-isotope labeled amino acid derivative used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is supplied as a white to off-white solid of high purity suitable for analytical and research applications.
  • Deuterium-labeled Fmoc-protected leucine for isotope-dilution quantitation.
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • High purity (≈99.3%) for accurate analytical results.
  • White to off-white solid form for easy handling and storage.
  • Molecular weight 356.43 and CAS number 538372-74-6.