Learn More
Medchemexpress LLC N-Fmoc-N,O-dimethyl-L-serine | 1569103-64-5 | MFCD31730320 | 97.9% | 355.38 g/mol | C20H21NO5 | 5 G
SDP
Catalog No. 5000301675
Encompass_Preferred
N-Fmoc-N,O-dimethyl-L-serine is an Fmoc-protected serine derivative used as a building block in peptide and organic synthesis. It is commonly used in medicinal chemistry research and has been reported as an intermediate in coibamide A synthesis. The material is supplied as a solid with available analytical documentation for laboratory use.
  • Fmoc-protected serine derivative for peptide and organic synthesis.
  • High purity suitable for research (typical purity ~97.9%).
  • Solid, white to light yellow appearance for easy handling.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Stable as a powder when stored at -20°C for extended periods.
  • Datasheet, certificate of analysis, and safety sheet available.
Catalog No. 50-003-01675 Supplier Medchemexpress LLC Supplier No. HYW0391025G
N-Fmoc-N,O-dimethyl-L-serine is an Fmoc-protected serine derivative used as a building block in peptide and organic synthesis. It is commonly used in medicinal chemistry research and has been reported as an intermediate in coibamide A synthesis. The material is supplied as a solid with available analytical documentation for laboratory use.
  • Fmoc-protected serine derivative for peptide and organic synthesis.
  • High purity suitable for research (typical purity ~97.9%).
  • Solid, white to light yellow appearance for easy handling.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Stable as a powder when stored at -20°C for extended periods.
  • Datasheet, certificate of analysis, and safety sheet available.