Learn More
Medchemexpress LLC N-Fmoc-O-benzyl-L-tyrosine | 71989-40-7 | MFCD00065682 | 99.2% | C31H27NO5 | 10 G
SDP
Catalog No. 5000434424
Encompass_Preferred
N-Fmoc-O-benzyl-L-tyrosine is a tyrosine derivative with a molecular weight of 493.55 and the chemical formula C31H27NO5. It appears as a white to off-white solid with a purity of 99.15%. This product is intended for research use only. Amino acids and their derivatives, such as N-Fmoc-O-benzyl-L-tyrosine, are commercially utilized as ergogenic supplements.
  • Influences the secretion of anabolic hormones
  • Provides fuel during exercise
  • Enhances mental performance during stress-related tasks
  • Helps prevent exercise-induced muscle damage
  • Recognized as a beneficial ergogenic dietary substance
  • Soluble in DMSO at a concentration of at least 30 mg/mL
  • For in vivo experiments, soluble in 10% DMSO and 90% corn oil at least 2.5 mg/mL
Catalog No. 50-004-34424 Supplier Medchemexpress LLC Supplier No. HY7913210G
N-Fmoc-O-benzyl-L-tyrosine is a tyrosine derivative with a molecular weight of 493.55 and the chemical formula C31H27NO5. It appears as a white to off-white solid with a purity of 99.15%. This product is intended for research use only. Amino acids and their derivatives, such as N-Fmoc-O-benzyl-L-tyrosine, are commercially utilized as ergogenic supplements.
  • Influences the secretion of anabolic hormones
  • Provides fuel during exercise
  • Enhances mental performance during stress-related tasks
  • Helps prevent exercise-induced muscle damage
  • Recognized as a beneficial ergogenic dietary substance
  • Soluble in DMSO at a concentration of at least 30 mg/mL
  • For in vivo experiments, soluble in 10% DMSO and 90% corn oil at least 2.5 mg/mL