Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-O-methyl-D-tyrosine, 98%

Catalog No. AAH6307303
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6307303 1 g
AAH6307306 5 g
AAH63073MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6307303 Supplier Thermo Scientific Chemicals Supplier No. H6307303
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 201335-88-8 | C25H23NO5 | 417.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201335-88-8
Molecular Formula C25H23NO5
Molecular Weight (g/mol) 417.46
MDL Number MFCD00237393
InChI Key JYQODLWFOPCSCS-UHFFFAOYNA-N
Synonym fmoc-d-tyr me-oh, fmoc-o-methyl-d-tyrosine, fmoc-d-4-methoxyphenylalanine, fmoc-4-methoxy-d-phenylalanine, fmoc-4-methoxy-d-phe-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-methoxyphenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-methoxyphenyl propanoic acid, fmoc-d-4-methoxy-phe-oh, fmoc-d-tyr me, fmoc-d-4-meo-phe-oh
PubChem CID 7019819
SMILES COC1=CC=C(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1

Specifications

Color White
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-methoxyphenyl)propanoic acid
Quantity 1 g
Formula Weight 417.45
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Fmoc-O-methyl-D-tyrosine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.