Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-O-tert-butyl-D-serine, 98%

Catalog No. AAH6243906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6243906 5 g
AAH6243914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6243906 Supplier Thermo Scientific Chemicals Supplier No. H6243906
  • $122.10
    $103.65 / Each of 1

    Save $18.45

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 128107-47-1 | C22H25NO5 | 383.444 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128107-47-1
Molecular Formula C22H25NO5
Molecular Weight (g/mol) 383.444
MDL Number MFCD00077071
InChI Key REITVGIIZHFVGU-LJQANCHMSA-N
Synonym fmoc-d-ser tbu-oh, fmoc-o-tert-butyl-d-serine, fmoc-d-ser but-oh, pubchem10040, ksc915q4f, o-tert-butyl-n-fmoc-l-serine, 2r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, d-serine, o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl, 2r-3-tert-butoxy-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, fmoc-o-tert.butyl-d-serine
PubChem CID 7004204
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]propanoic acid
SMILES CC(C)(C)OCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 131°C
Quantity 5 g
Beilstein 5309984
Formula Weight 383.44
Percent Purity 98%
Chemical Name or Material N-Fmoc-O-tert-butyl-D-serine
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.