Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-O-tert-butyl-D-tyrosine, 95%

Catalog No. AAH6276503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6276503 1 g
AAH6276506 5 g
AAH6276514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6276503 Supplier Thermo Scientific Chemicals Supplier No. H6276503
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 118488-18-9 | C28H29NO5 | 459.54 g/mol

N-Fmoc-O-tert-butyl-D-tyrosine is potentially useful for proteomics studies and solid phase peptide synthesis techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-O-tert-butyl-D-tyrosine is potentially useful for proteomics studies and solid phase peptide synthesis techniques.

Solubility
Soluble in 1 mmole in 2 ml dimethylformamide (Clearly soluble). Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118488-18-9
Molecular Formula C28H29NO5
Molecular Weight (g/mol) 459.54
MDL Number MFCD00065684
InChI Key JAUKCFULLJFBFN-KSYWNVGFNA-N
Synonym fmoc-d-tyr tbu-oh, fmoc-o-tert-butyl-d-tyrosine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, nalpha-9h-fluoren-9-ylmethoxy carbonyl-o-tert-butyl-d-tyrosine, 2r-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, d-tyrosine, o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl, ambotzfaa1341, pubchem10054, fmoc-d-tyr but-oh, fmoc-o-tert.butyl-d-tyrosine
PubChem CID 12968135
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid
SMILES CC(C)(C)OC1=CC=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1

Specifications

Quantity 1 g
Beilstein 6691868
Solubility Information Soluble in 1 mmole in 2 ml dimethylformamide (Clearly soluble). Slightly soluble in water.
Formula Weight 459.53
Percent Purity 95%
Chemical Name or Material N-Fmoc-O-tert-butyl-D-tyrosine
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.