Learn More
Abcam N-FMOC Rhodamine 110, 10MG
Catalog No. 50996107
Encompass_Preferred
MW 552.6 Da, Purity >98%. Green fluorophore used in enyme assay systems. Mono-protected rhodamine 110 derivative useful for the synthesis of heterofunctional R110 derivatives, including peptidase substrates. The FMOC protection group is common in many standard peptide synthesis systems. Free R110 (e=88K).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-107 Supplier Abcam Supplier No. AB27591710MG
MW 552.6 Da, Purity >98%. Green fluorophore used in enyme assay systems. Mono-protected rhodamine 110 derivative useful for the synthesis of heterofunctional R110 derivatives, including peptidase substrates. The FMOC protection group is common in many standard peptide synthesis systems. Free R110 (e=88K).
The product is subject to the following: Abcam Restricted Use Statement