Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-S-(4-methoxybenzyl)-L-cysteine, 95%, Thermo Scientific™

Catalog No. AAH6304506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6304506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6304506 Supplier Thermo Scientific Chemicals Supplier No. H6304506

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141892-41-3
Molecular Formula C26H25NO5S
Molecular Weight (g/mol) 463.548
MDL Number MFCD00065635
InChI Key IWZGYHFOLFRYPK-DEOSSOPVSA-N
Synonym fmoc-cys pmeobzl-oh, fmoc-s-p-methoxybenzyl-l-cysteine, fmoc-cys mob-oh, n-fmoc-s-4-methoxybenzyl-l-cysteine, fmoc-s-4-methoxybenzyl-l-cysteine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-methoxybenzyl thio propanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-s-4-methoxybenzyl-l-cysteine, fmoc-cys 4-meobzl-oh, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, pubchem19018
PubChem CID 11236605
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(4-methoxyphenyl)methylsulfanyl]propanoic acid
SMILES COC1=CC=C(C=C1)CSCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Color White
Quantity 5 g
Formula Weight 463.55
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-S-(4-methoxybenzyl)-L-cysteine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.