Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-S-benzyl-L-cysteine, 98%, Thermo Scientific™

Catalog No. AAH6302806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6302806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6302806 Supplier Thermo Scientific Chemicals Supplier No. H6302806

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53298-33-2
Molecular Formula C25H23NO4S
Molecular Weight (g/mol) 433.522
MDL Number MFCD00065633
InChI Key AKXYVGAAQGLAMD-QHCPKHFHSA-N
Synonym fmoc-cys bzl-oh, fmoc-s-benzyl-l-cysteine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzylthio propanoic acid, l-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-phenylmethyl, 2r-3-benzylsulfanyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-s-benzyl-l-cys, pubchem12391, fmoc-l-cys bzl-oh, ksc925c9h, n-fmoc-s-phenylmethyl-l-cysteine
PubChem CID 11750859
IUPAC Name (2R)-3-benzylsulfanyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)CSCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 5 g
Formula Weight 433.52
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-S-benzyl-L-cysteine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.