Learn More
Cayman Chemical N-formyl-2-Aminophenol, Purity: ≥98%, CAS: 2843-27-8, 100 mg
SDP
Catalog No. 502909832
Encompass_Preferred
A bacterial secondary metabolite with antioxidant activity; scavenges DPPH radicals in a cell-free assay (IC50 = 3.23 µg/ml); has been used in the synthesis of compounds with antibacterial and antifungal activities
Catalog No. 50-290-9832 Supplier Cayman Chemical Supplier No. 36802100 MG
A bacterial secondary metabolite with antioxidant activity; scavenges DPPH radicals in a cell-free assay (IC50 = 3.23 µg/ml); has been used in the synthesis of compounds with antibacterial and antifungal activities