Learn More
Cayman Chemical N-formyl 4-ANPP, Purity: ≥98%, CAS: 66670-15-3, 1 mg
SDP
Catalog No. 502918320
Encompass_Preferred
An analytical reference standard that is structurally similar to known opioids; intended for research and forensic applications
Catalog No. 50-291-8320 Supplier Cayman Chemical Supplier No. 388631 MG
An analytical reference standard that is structurally similar to known opioids; intended for research and forensic applications