Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Formyl-L-leucine, TCI America™
SDP

Catalog No. F01311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F01311G Supplier TCI America Supplier No. F01311G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6113-61-7
Molecular Formula C7H13NO3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00055861
InChI Key HFBHOAHFRNLZGN-MDOHGIEYNA-N
Synonym n-formyl-l-leucine, n-formyl-leu-oh, n-formylleucine, l-leucine, n-formyl, unii-5tha63y4ff, 5tha63y4ff, 2s-2-formamido-4-methylpentanoic acid, n-formylleucine #, formylleucin, n-formyl leucine
PubChem CID 9880216
IUPAC Name (2S)-2-formamido-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC=O)C(O)=O

Specifications

Melting Point 142°C
Color White
Quantity 1 g
Formula Weight 159.19
Physical Form Crystalline Powder
Chemical Name or Material N-Formyl-L-leucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.