Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Formyl-L-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. F01271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F01271G Supplier TCI America Supplier No. F01271G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13200-85-6
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD00021030
InChI Key NSTPXGARCQOSAU-VIFPVBQESA-N
PubChem CID 759256
IUPAC Name (2S)-2-formamido-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)NC=O

Specifications

Melting Point 165°C
Color White
Quantity 1 g
Formula Weight 193.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Formyl-L-phenylalanine
EINECSNumber (3)-4113
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.