Learn More
Medchemexpress LLC N-Formyl-Met-Ala-Ser | 17351-32-5 | MFCD00134891 | 98.1% | 335.38 | 5 MG
SDP
Catalog No. 5000423708
Encompass_Preferred
N-Formyl-Met-Ala-Ser is a peptide that binds to formyl peptide receptors on neutrophils. It is presented as a white to off-white solid. This compound is suitable for various laboratory applications, particularly in the study of inflammation and the immune system.
  • Peptide that binds to formyl peptide receptors on neutrophils
  • Presented as a white to off-white solid
  • Suitable for inflammation and immunology research
Catalog No. 50-004-23708 Supplier Medchemexpress LLC Supplier No. HYP17565MG
N-Formyl-Met-Ala-Ser is a peptide that binds to formyl peptide receptors on neutrophils. It is presented as a white to off-white solid. This compound is suitable for various laboratory applications, particularly in the study of inflammation and the immune system.
  • Peptide that binds to formyl peptide receptors on neutrophils
  • Presented as a white to off-white solid
  • Suitable for inflammation and immunology research