Learn More
Medchemexpress LLC N-Formyl-MMYALF | 863418-59-1 | 99.8% | 803.00 | 1 MG
SDP
Catalog No. 5000425218
Encompass_Preferred
N-Formyl-MMYALF is a potent mitochondrial N-formyl peptide (mtFP) that depletes calcium ions in the endoplasmic reticulum. It can inhibit the FPR-1-mediated chemotactic response of polymorphonuclear leukocytes (PMNs) to bacterial peptides.
  • Potent mitochondrial N-formyl peptide
  • Depletes calcium ions in the endoplasmic reticulum
  • Inhibits chemotactic response of polymorphonuclear leukocytes
  • Used as an inhibitor or substrate
  • Applicable in infection and bacterial infection research
Catalog No. 50-004-25218 Supplier Medchemexpress LLC Supplier No. HYP107381MG
N-Formyl-MMYALF is a potent mitochondrial N-formyl peptide (mtFP) that depletes calcium ions in the endoplasmic reticulum. It can inhibit the FPR-1-mediated chemotactic response of polymorphonuclear leukocytes (PMNs) to bacterial peptides.
  • Potent mitochondrial N-formyl peptide
  • Depletes calcium ions in the endoplasmic reticulum
  • Inhibits chemotactic response of polymorphonuclear leukocytes
  • Used as an inhibitor or substrate
  • Applicable in infection and bacterial infection research