Learn More
Medchemexpress LLC N-formylmethionine | 4289-98-9 | 99.9% | 177.22 g/mol | C6H11NO3S | 100 MG
SDP
Catalog No. 5000301329
Encompass_Preferred
For-Met-OH (N-formylmethionine) is an endogenous metabolite used as an analytical standard and research reagent. It is involved in translation initiation in bacteria, mitochondria, and chloroplasts and is investigated as a potential biomarker in metabolic and critical-illness studies. Available in both solid and solution forms for analytical and biological workflows.
  • High purity, typically 99.92%.
  • Available as solid (100 mg) and solution (10 mM in DMSO) packs.
  • Suitable as an analytical standard for metabolomics and biomarker studies.
  • Compact molecular weight of 177.22 g/mol for mass spectrometry applications.
  • Datasheet and certificate of analysis available for quality assurance.
Catalog No. 50-003-01329 Supplier Medchemexpress LLC Supplier No. HYW016647100MG
For-Met-OH (N-formylmethionine) is an endogenous metabolite used as an analytical standard and research reagent. It is involved in translation initiation in bacteria, mitochondria, and chloroplasts and is investigated as a potential biomarker in metabolic and critical-illness studies. Available in both solid and solution forms for analytical and biological workflows.
  • High purity, typically 99.92%.
  • Available as solid (100 mg) and solution (10 mM in DMSO) packs.
  • Suitable as an analytical standard for metabolomics and biomarker studies.
  • Compact molecular weight of 177.22 g/mol for mass spectrometry applications.
  • Datasheet and certificate of analysis available for quality assurance.