Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Formylmethionine Specific Transfer Ribonucleic From E. coli MRE 600, MP Biomedicals™

Catalog No. MP219915410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219915410 10 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219915410 Supplier MP Biomedicals, Inc Supplier No. 0219915410

missing translation for 'validMainframeMsg'

N-Formylmethionine Specific Transfer Ribonucleic From E. coli MRE 600 Activity=1300 ± 50 picomoles per A260 units, White Powder

  • Ribonucleic acid, transfer, N-formylmethionine-specific may be used as a substrate to study the specifity and kinetics of natural, mutated and chemically engineered N-formylmethionine-specific E. coli aminoacyl-tRNA synthetase(s) and methionyl-tRNA formyltransferase(s) (MTF).
  • Each T-RNA molecule is specific for and binds with a particular amino acid; more than one exists for each amino acid.
  • T-RNA performs three functions during protein synthesis: binds with its specific amino acid, recognizes the corresponding codon on mRNA and places the amino acid in the correct position for attachment to the polypeptide chain being formed & binds the polypeptide to the ribosome.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-63-0
MDL Number MFCD00132196
IUPAC Name Ribonucleic acid

Specifications

Color White
Quantity 10 U
Physical Form Powder
RTECSNumber RF0772520
storageNote1 -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.