Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Formylsaccharin 98.0+%, TCI America™
SDP

Catalog No. F085425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F085425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F085425G Supplier TCI America Supplier No. F085425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50978-45-5
Molecular Formula C8H5NO4S
Molecular Weight (g/mol) 211.19
MDL Number MFCD23380208
InChI Key LMPIFZSJKLLOLM-UHFFFAOYSA-N
PubChem CID 54671689
IUPAC Name 1,1,3-trioxo-2,3-dihydro-1λ⁶,2-benzothiazole-2-carbaldehyde
SMILES O=CN1C(=O)C2=CC=CC=C2S1(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 211.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Formylsaccharin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.