Learn More
Medchemexpress LLC N-(furan-2-ylmethyl)-3-[4-[methyl(propyl)amino]-6-(trifluoromethyl)pyrimidin-2-yl]sulfanylpropanamid | 2313525-20-9 | 99.8% | 402.43 g/mol | C17H21F3N4O2S | 5 MG
SDP
Catalog No. 5000262128
Encompass_Preferred
SET2 is a small-molecule selective TRPV2 antagonist used for laboratory research. It inhibits TRPV2 with an IC50 of 0.46 μM and has been shown to block TRP channel activity and suppress migration of prostate cancer cells. Supplied as a white to off-white solid of high purity, it is intended for research use only.
  • Selective TRPV2 antagonist with IC50 = 0.46 μM.
  • High purity (~99.8%) suitable for biochemical studies.
  • Solid form, easy to weigh and handle.
  • Reduces lysophosphatidic acid-induced cytoplasmic calcium increases.
  • Demonstrated suppression of prostate cancer cell migration in cellular assays.
  • Recommended storage: 4°C; in solvent: -80°C for long-term stability.
Catalog No. 50-002-62128 Supplier Medchemexpress LLC Supplier No. HY1322225MG
SET2 is a small-molecule selective TRPV2 antagonist used for laboratory research. It inhibits TRPV2 with an IC50 of 0.46 μM and has been shown to block TRP channel activity and suppress migration of prostate cancer cells. Supplied as a white to off-white solid of high purity, it is intended for research use only.
  • Selective TRPV2 antagonist with IC50 = 0.46 μM.
  • High purity (~99.8%) suitable for biochemical studies.
  • Solid form, easy to weigh and handle.
  • Reduces lysophosphatidic acid-induced cytoplasmic calcium increases.
  • Demonstrated suppression of prostate cancer cell migration in cellular assays.
  • Recommended storage: 4°C; in solvent: -80°C for long-term stability.