Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N(G)-monomethyl-L-arginine acetate | 53308-83-1 | MFCD00069311 | 99.0% | 248.28 g/mol | C9H20N4O4 | 10 MG
SDP

Catalog No. 5000295332
Encompass_Preferred

L-NMMA acetate (N(G)-monomethyl-L-arginine acetate) is a small-molecule nitric oxide synthase (NOS) inhibitor supplied as a white to off-white solid for laboratory research. It is used in biochemical assays and NOS inhibition studies and is provided in milligram-scale quantities for analytical and experimental use.

  • Non-selective nitric oxide synthase inhibitor effective against nNOS, iNOS, and eNOS.
  • High purity: 99.0% as stated by manufacturer.
  • Molecular weight 248.28 g/mol, molecular formula C9H20N4O4.
  • White to off-white solid form suitable for laboratory handling and storage.
  • Available in small pack sizes (e.g., 1 mg, 5 mg, 10 mg) for research applications.
  • Commonly used as an analytical standard and biochemical reagent in NOS-related studies.

Catalog No. 50-002-95332 Supplier Medchemexpress LLC Supplier No. HY18732A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-NMMA acetate (N(G)-monomethyl-L-arginine acetate) is a small-molecule nitric oxide synthase (NOS) inhibitor supplied as a white to off-white solid for laboratory research. It is used in biochemical assays and NOS inhibition studies and is provided in milligram-scale quantities for analytical and experimental use.

  • Non-selective nitric oxide synthase inhibitor effective against nNOS, iNOS, and eNOS.
  • High purity: 99.0% as stated by manufacturer.
  • Molecular weight 248.28 g/mol, molecular formula C9H20N4O4.
  • White to off-white solid form suitable for laboratory handling and storage.
  • Available in small pack sizes (e.g., 1 mg, 5 mg, 10 mg) for research applications.
  • Commonly used as an analytical standard and biochemical reagent in NOS-related studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.