Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(gamma-Maleimidobutyryloxy)succinimide

Catalog No. AAJ61850MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61850MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61850MC Supplier Thermo Scientific Chemicals Supplier No. J61850MC
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 80307-12-6 | C12H12N2O6 | 280.24 g/mol

A heterobifunctional cross-linking reagent with amine and sulfhydryl reactivity. Useful in enzyme immunoassays

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A heterobifunctional cross-linking reagent with amine and sulfhydryl reactivity
  • Useful in enzyme immunoassays
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80307-12-6
Molecular Formula C12H12N2O6
Molecular Weight (g/mol) 280.24
MDL Number MFCD00036817
InChI Key PVGATNRYUYNBHO-UHFFFAOYSA-N
Synonym gmbs, 4-maleimidobutyric acid n-hydroxysuccinimide ester, 4-maleimidobutyric acid n-succinimidyl ester, n-gamma-maleimidobutyryloxy succinimide, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl butanoate, n-4-maleimidobutyryloxy succinimide, n-succinimidyl 4-maleimidobutyrate, n-succinimidyl-4-maleimidobutyrate, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxopyrrol-1-yl butanoate
PubChem CID 133440
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-(2,5-dioxopyrrol-1-yl)butanoate
SMILES O=C(CCCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O

Specifications

Melting Point 130°C to 132°C
Color White
Quantity 100 mg
Beilstein 6427689
Formula Weight 280.23
Physical Form Powder
Chemical Name or Material N-(gamma-Maleimidobutyryloxy)succinimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.