Learn More
American Research Products Inc N-Glycan Rapid Detection Kit
Catalog No. 5000803916
Encompass
PNGase F cleavage of N-glycan and 2-AB labeling andpurification of monoclonal antibodies and other glycoproteins(based on magnetic bead purification 25/50 rxns/kit) This kit supports high-throughput operation of a 96-well plate and supports two N-glycan preparation methods non-denaturing (for common monoclonal antibodies) anddenaturing (for difficult-to-cleave glycoproteins) The denaturingmethod is more universal
Catalog No. 50-008-03916 Supplier American Research Products Inc Supplier No. ARPERNG02
PNGase F cleavage of N-glycan and 2-AB labeling andpurification of monoclonal antibodies and other glycoproteins(based on magnetic bead purification 25/50 rxns/kit) This kit supports high-throughput operation of a 96-well plate and supports two N-glycan preparation methods non-denaturing (for common monoclonal antibodies) anddenaturing (for difficult-to-cleave glycoproteins) The denaturingmethod is more universal