Learn More
Agilent Technologies N-GLYCANASE PLUS 400MU
Supplier:  Agilent Technologies GKE5010B
Encompass
PNGase F, peptide-N-glycosidase F,peptide-N4-(N-acetyl--glucosaminyl)asparagine amidase.Releases intact N-glycans by cleavingbetween the innermost GlcNAc and Asn.
Catalog No. NC9801615