Learn More
TARGETMOL CHEMICALS INC N-Glycolylneuraminic acid 10MG
SDP
Catalog No. 502878382
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. N-Glycolylneuraminic acid (GcNeu) is a nonhuman sialic acid molecule synthesized in pigs. N-Glycolylneuraminic acid is a receptor of human and animal IAVs. Purity ≥98%
Catalog No. 50-287-8382 Supplier TARGETMOL CHEMICALS INC Supplier No. T1945610MG
Add to Cart
Add to Cart
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. N-Glycolylneuraminic acid (GcNeu) is a nonhuman sialic acid molecule synthesized in pigs. N-Glycolylneuraminic acid is a receptor of human and animal IAVs. Purity ≥98%