Learn More
Medchemexpress LLC N-Glycolylneuraminic acid | 1113-83-3 | 99.8% | 10 MG
SDP
Catalog No. 5000340854
Encompass_Preferred
N-Glycolylneuraminic acid is a nonhuman sialic acid molecule, synthesized in pigs but not in humans. It functions as a decoy receptor for N-Glycolylneuraminic acid-binding influenza A viruses (IAVs), making it suitable for research applications.
  • Nonhuman sialic acid molecule
  • Functions as a decoy receptor for influenza A viruses
  • Prevents influenza virus entry into A549 cells in vitro studies
  • Available as a white to off-white solid
  • For research use only
Catalog No. 50-003-40854 Supplier Medchemexpress LLC Supplier No. HY12896510MG
N-Glycolylneuraminic acid is a nonhuman sialic acid molecule, synthesized in pigs but not in humans. It functions as a decoy receptor for N-Glycolylneuraminic acid-binding influenza A viruses (IAVs), making it suitable for research applications.
  • Nonhuman sialic acid molecule
  • Functions as a decoy receptor for influenza A viruses
  • Prevents influenza virus entry into A549 cells in vitro studies
  • Available as a white to off-white solid
  • For research use only