Learn More
Cayman Chemical N-GlycolylnuramInIc AcId 50mg
SDP
Catalog No. 502992341
Encompass_Preferred
A sialic acid; not endogenously produced in humans but accumulates in human cells after exogenous ingestion from dietary sources; accumulates in human tumors and uptake is enhanced under hypoxic conditions
Catalog No. 50-299-2341 Supplier Cayman Chemical Supplier No. 3028350 MG
A sialic acid; not endogenously produced in humans but accumulates in human cells after exogenous ingestion from dietary sources; accumulates in human tumors and uptake is enhanced under hypoxic conditions