Learn More
New England Biolabs, Inc. N-Glycopeptide Binding Protein - 1 ml
SDP
Catalog No. 502581805
Encompass
N-Glycopeptide Binding Protein, also known as Fbs1-GYR lectin, is engineered to selectively bind and enrich a wide variety of N-glycopeptides in a complex proteomic sample to assist N-glycoprotein identification and discovery, without requiring the use of traditional lectin affinity chromatography (LAC) techniques, HILIC columns or cartridges. This engineered lectin, originating from human Fbs1 protein, exhibits improved specificity towards high mannose, complex, and hybrid N-glycans, as well as towards fucosylated and non-fucosylated, sialylated and non-sialylated, with bi-, tri- and tetra-antennary structures. N-Glycopeptide Binding Protein includes a SNAP tag for immobilizing to create versatile workflows. 1 mL of protein at 1 mg/mL Source: An E. coli strain that carries human Fbs1 mutant gene. Storage Buffer: 20 mM Tris-HCl, 50 mM NaCl, 0.2 mM TCEP, pH 8.0 Heat Inactivation: No
Catalog No. 50-258-1805 Supplier New England Biolabs, Inc. Supplier No. P0872S
N-Glycopeptide Binding Protein, also known as Fbs1-GYR lectin, is engineered to selectively bind and enrich a wide variety of N-glycopeptides in a complex proteomic sample to assist N-glycoprotein identification and discovery, without requiring the use of traditional lectin affinity chromatography (LAC) techniques, HILIC columns or cartridges. This engineered lectin, originating from human Fbs1 protein, exhibits improved specificity towards high mannose, complex, and hybrid N-glycans, as well as towards fucosylated and non-fucosylated, sialylated and non-sialylated, with bi-, tri- and tetra-antennary structures. N-Glycopeptide Binding Protein includes a SNAP tag for immobilizing to create versatile workflows. 1 mL of protein at 1 mg/mL Source: An E. coli strain that carries human Fbs1 mutant gene. Storage Buffer: 20 mM Tris-HCl, 50 mM NaCl, 0.2 mM TCEP, pH 8.0 Heat Inactivation: No