Learn More
Enzo Life Sciences N. gonorrhoeae native antigen (1ml)
SDP
Supplier:  Enzo Life Sciences ALX6301291000
Encompass_Preferred
Alternative name: Neisseria gonorrhoeae native antigen. Purity: ≥99%. Formulation: Liquid. In PBS. Source: From Neisseria gonorrhoeae, strain ATCC 19424. Inactivated by 3 hours at 56°C. Long Term Storage: -80°C. Applications: ELISA LINE assay, Immunogen, Cytotoxicity studies, Antibody capture.
Catalog No. 50-200-9654