Learn More
Medchemexpress LLC N-heptadecane | 629-78-7 | MFCD00009002 | 97.2% | 240.47 g/mol | C17H36 | 1 KG
SDP
Catalog No. 5000282995
Encompass_Preferred
N-Heptadecane (heptadecane) is a straight-chain saturated hydrocarbon (C17H36) used as a reagent and analytical standard in research and analytical laboratories. It is supplied for laboratory use only and is available in multiple pack sizes, including a 1 kg package.
  • Straight-chain alkane suitable as an analytical standard.
  • High purity, 97.2%, for consistent analytical results.
  • Nonpolar and hydrophobic, useful in chromatography and method development.
  • Available in multiple pack sizes to support both small-scale and bulk use.
  • Provided with SDS and COA documentation for handling and quality verification.
Catalog No. 50-002-82995 Supplier Medchemexpress LLC Supplier No. HYW1313511KG
N-Heptadecane (heptadecane) is a straight-chain saturated hydrocarbon (C17H36) used as a reagent and analytical standard in research and analytical laboratories. It is supplied for laboratory use only and is available in multiple pack sizes, including a 1 kg package.
  • Straight-chain alkane suitable as an analytical standard.
  • High purity, 97.2%, for consistent analytical results.
  • Nonpolar and hydrophobic, useful in chromatography and method development.
  • Available in multiple pack sizes to support both small-scale and bulk use.
  • Provided with SDS and COA documentation for handling and quality verification.