Learn More
Medchemexpress LLC N-heptadecane | 629-78-7 | MFCD00009002 | 97.2% | 240.47 g/mol | C17H36 | 10 G
SDP
Catalog No. 5000282994
Encompass_Preferred
N-Heptadecane (C17H36) is a straight-chain C17 alkane used as an analytical standard and reference material in chemical analysis. It is suitable for chromatographic calibration, method development, and use as an internal standard or tracer. Stability, purity, and storage instructions are provided in the product documentation.
  • Provides a stable C17 hydrocarbon reference for chromatographic calibration.
  • Suitable as an internal standard or tracer in analytical assays.
  • High purity with certificate of analysis for traceability.
  • Stable under recommended storage conditions for long-term use.
  • Available in small laboratory quantities for analytical workflows.
Catalog No. 50-002-82994 Supplier Medchemexpress LLC Supplier No. HYW13135110G
N-Heptadecane (C17H36) is a straight-chain C17 alkane used as an analytical standard and reference material in chemical analysis. It is suitable for chromatographic calibration, method development, and use as an internal standard or tracer. Stability, purity, and storage instructions are provided in the product documentation.
  • Provides a stable C17 hydrocarbon reference for chromatographic calibration.
  • Suitable as an internal standard or tracer in analytical assays.
  • High purity with certificate of analysis for traceability.
  • Stable under recommended storage conditions for long-term use.
  • Available in small laboratory quantities for analytical workflows.