Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-heptadecane | 629-78-7 | MFCD00009002 | 97.2% | 240.47 g/mol | C17H36 | 50 G
SDP

Catalog No. 5000283307
Encompass_Preferred

N-Heptadecane is a straight-chain saturated hydrocarbon used as a technical reagent and analytical standard. It serves as an internal standard, tracer, and reference compound in chromatographic and mass spectrometric analyses, and is supplied with supporting documentation for laboratory use.

  • High purity analytical reagent suitable for standards and calibration.
  • Useful as an internal standard or tracer in GC/MS and chromatographic methods.
  • Chemical formula C17H36; molecular weight 240.47 g/mol.
  • Available in laboratory pack sizes including 50 G.
  • Storage recommendations: pure form at -20°C (long-term) or 4°C (short-term); in solvent at -80°C for extended stability.
  • Documentation available: data sheet, SDS, COA.

Catalog No. 50-002-83307 Supplier Medchemexpress LLC Supplier No. HYW13135150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Heptadecane is a straight-chain saturated hydrocarbon used as a technical reagent and analytical standard. It serves as an internal standard, tracer, and reference compound in chromatographic and mass spectrometric analyses, and is supplied with supporting documentation for laboratory use.

  • High purity analytical reagent suitable for standards and calibration.
  • Useful as an internal standard or tracer in GC/MS and chromatographic methods.
  • Chemical formula C17H36; molecular weight 240.47 g/mol.
  • Available in laboratory pack sizes including 50 G.
  • Storage recommendations: pure form at -20°C (long-term) or 4°C (short-term); in solvent at -80°C for extended stability.
  • Documentation available: data sheet, SDS, COA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.