Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-heptadecane | 629-78-7 | MFCD00009002 | 97.2% | 240.5 g/mol | C17H36 | 25 G
SDP

Catalog No. 5000283405
Encompass_Preferred

N-Heptadecane is a straight-chain C17 alkane provided as an analytical standard for research and analytical applications. It is commonly used as a reference compound for chromatographic calibration, retention time markers, and analytical method development. The product is supplied with documented quality information and is available in multiple pack sizes to suit laboratory needs.

  • Straight-chain C17 alkane suitable for analytical use.
  • Purity 97.2%.
  • CAS number 629-78-7.
  • Supplied with data sheet, SDS, and COA.
  • Available in multiple pack sizes including 25 g.
  • Useful for chromatographic calibration and retention time markers.
  • Suitable for reference standard preparation and method development.

Catalog No. 50-002-83405 Supplier Medchemexpress LLC Supplier No. HYW13135125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Heptadecane is a straight-chain C17 alkane provided as an analytical standard for research and analytical applications. It is commonly used as a reference compound for chromatographic calibration, retention time markers, and analytical method development. The product is supplied with documented quality information and is available in multiple pack sizes to suit laboratory needs.

  • Straight-chain C17 alkane suitable for analytical use.
  • Purity 97.2%.
  • CAS number 629-78-7.
  • Supplied with data sheet, SDS, and COA.
  • Available in multiple pack sizes including 25 g.
  • Useful for chromatographic calibration and retention time markers.
  • Suitable for reference standard preparation and method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.