Learn More
Medchemexpress LLC N-Heptadecanoyl-D-erythro-sphingosine | 67492-16-4 | MFCD00274182 | 98.0% | 551.93 | C35H69NO3 | 5 MG
SDP
Catalog No. 5000264033
Encompass_Preferred
C17 ceramide (d18:1/17:0) is a synthetic ceramide provided as a high-purity white to off-white solid for biochemical and cell biology research. It is commonly used to investigate lipid signaling, membrane properties, and apoptosis pathways, and can be formulated for in vitro assays and analytical studies.
  • High purity (98.0%) suitable for analytical and biological assays.
  • Molecular weight 551.93 and formula C35H69NO3 for accurate calculations.
  • Supplied as a solid powder for ease of handling and storage.
  • Available in small research pack sizes for low-volume experiments.
  • Powder stable at -20°C for long-term storage.
  • Recommended solvent storage at -80°C for prolonged sample preservation.
Catalog No. 50-002-64033 Supplier Medchemexpress LLC Supplier No. HY1157035MG
C17 ceramide (d18:1/17:0) is a synthetic ceramide provided as a high-purity white to off-white solid for biochemical and cell biology research. It is commonly used to investigate lipid signaling, membrane properties, and apoptosis pathways, and can be formulated for in vitro assays and analytical studies.
  • High purity (98.0%) suitable for analytical and biological assays.
  • Molecular weight 551.93 and formula C35H69NO3 for accurate calculations.
  • Supplied as a solid powder for ease of handling and storage.
  • Available in small research pack sizes for low-volume experiments.
  • Powder stable at -20°C for long-term storage.
  • Recommended solvent storage at -80°C for prolonged sample preservation.