Learn More
Medchemexpress LLC N-heptadecanoyl-d-erythro-sphingosylphosphorylcholine | 121999-64-2 | 99.7% | 717.05 g/mol | C40H81N2O6P | 5 MG
SDP
Catalog No. 5000262815
Encompass_Preferred
C17 sphingomyelin (d18:1/17:0) is a synthetic sphingomyelin analog used as an internal standard for quantitative lipid analysis, particularly mass spectrometry-based sphingomyelin quantification. It is provided as a high-purity analytical-grade solid suitable for LC-MS workflows.
  • High purity (99.67%) for reliable quantitation.
  • Molecular formula C40H81N2O6P; molecular weight 717.05 g/mol.
  • Suitable as an internal standard for LC-MS quantitation of sphingomyelin.
  • Available in small pack sizes for analytical use.
Catalog No. 50-002-62815 Supplier Medchemexpress LLC Supplier No. HY1316765MG
C17 sphingomyelin (d18:1/17:0) is a synthetic sphingomyelin analog used as an internal standard for quantitative lipid analysis, particularly mass spectrometry-based sphingomyelin quantification. It is provided as a high-purity analytical-grade solid suitable for LC-MS workflows.
  • High purity (99.67%) for reliable quantitation.
  • Molecular formula C40H81N2O6P; molecular weight 717.05 g/mol.
  • Suitable as an internal standard for LC-MS quantitation of sphingomyelin.
  • Available in small pack sizes for analytical use.