Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Heptafluorobutyrylimidazole

Catalog No. 5566455G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-664-55G 5 g
55-664-51G 1 g
55-664-550G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 55-664-55G Supplier Sigma Aldrich Supplier No. 5566455G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • May form a precipitate and may darken on storage-will not effect performance.n-heptafluorobutyrylimidazole may be used as a derivatization reagent in the gas-liquid chromatographic (glc) analysis of the following compounds:
  • Deoxynivalenol in wheat
  • Antibiotics (gentamicin, tobramycin, netilmicin, and amikacin) in human serum
  • Melatonin
  • Heptafluorobutyryl compounds formed by acylation of indolealkylamines
  • Acylation products of of morphine, codeine and nalorphine

Specifications

CAS 32477-35-3
Density 1.490 g/mL (at 25°C (literature))
Boiling Point 161°C (lit.)
Molecular Formula C7H3F7N2O
Refractive Index n20/D 1.3865 (literature)
Linear Formula C7H3F7N2O
MDL Number MFCD00014503
Quantity 5 g
Synonym 1-(Perfluorobutyryl)imidazole; NSC 151966
Molecular Weight (g/mol) 264.1
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.