Learn More
AdipoGen N-Heptafluorobutyrylimidazole
SDP
Supplier:  AdipoGen CDXH0101G005
Encompass_Preferred
Chemical. CAS 32477-35-3. Formula C7H3F7N2O. MW 264.1. Synthetic Mild amine-group derivatizing reagent. Non-acidic by-product prevents decomposition and reduces GC column degradation.
Catalog No. 50-256-0735