Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-heptanoylglycine-d2 | 3008541-77-0 | 95.0% | 189.25 | C9H15D2NO3 | 1 MG
SDP

Catalog No. 5000283545
Encompass_Preferred

N-Heptanoylglycine-d2 is the deuterium-labeled analogue of N-heptanoylglycine used as a stable isotope internal standard and tracer in analytical chemistry. It is a solid (white to off-white) compound with formula C9H15D2NO3 and molecular weight 189.25, supplied at high purity for quantitative NMR, GC-MS, and LC-MS studies.

  • High isotopic labeling (d2) for reliable internal standard performance.
  • Suitable for use as tracer or internal standard in NMR, GC-MS, and LC-MS.
  • Solid, white to off-white appearance facilitating handling.
  • Purity 95.0% to support quantitative analyses.
  • Storage recommendations provided for long-term and in-solvent conditions.

Catalog No. 50-002-83545 Supplier Medchemexpress LLC Supplier No. HYW141927S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Heptanoylglycine-d2 is the deuterium-labeled analogue of N-heptanoylglycine used as a stable isotope internal standard and tracer in analytical chemistry. It is a solid (white to off-white) compound with formula C9H15D2NO3 and molecular weight 189.25, supplied at high purity for quantitative NMR, GC-MS, and LC-MS studies.

  • High isotopic labeling (d2) for reliable internal standard performance.
  • Suitable for use as tracer or internal standard in NMR, GC-MS, and LC-MS.
  • Solid, white to off-white appearance facilitating handling.
  • Purity 95.0% to support quantitative analyses.
  • Storage recommendations provided for long-term and in-solvent conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.