Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-hexacosane-d54 | 1219803-91-4 | 99.5% | 421.04 g/mol | C26D54 | 10 MG
SDP

Catalog No. 5000283269
Encompass_Preferred

N-Hexacosane-d54 is a deuterium-labeled N-hexacosane supplied as a solid tracer and internal standard for quantitative analysis in research applications. It is suitable for use in NMR, GC-MS, and LC-MS workflows where high isotopic enrichment and purity are required.

  • Isotopically labeled with deuterium (d54)
  • High purity: 99.5%
  • Formula C26D54; molecular weight 421.04 g/mol
  • White to off-white solid form
  • Suitable as a tracer or internal standard for NMR, GC-MS, and LC-MS
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years)

Catalog No. 50-002-83269 Supplier Medchemexpress LLC Supplier No. HYW131352S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Hexacosane-d54 is a deuterium-labeled N-hexacosane supplied as a solid tracer and internal standard for quantitative analysis in research applications. It is suitable for use in NMR, GC-MS, and LC-MS workflows where high isotopic enrichment and purity are required.

  • Isotopically labeled with deuterium (d54)
  • High purity: 99.5%
  • Formula C26D54; molecular weight 421.04 g/mol
  • White to off-white solid form
  • Suitable as a tracer or internal standard for NMR, GC-MS, and LC-MS
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.