Learn More
Medchemexpress LLC N-hexacosane-d54 | 1219803-91-4 | MFCD03428117 | 99.5% | 421.04 g/mol | C26D54 | 5 MG
SDP
Catalog No. 5000283308
Encompass_Preferred
N-hexacosane-d54 is a deuterium-labeled analog of n-hexacosane designed for research applications as a tracer and internal standard in quantitative analyses such as NMR, GC-MS, and LC-MS. It is supplied as a solid with high isotopic and chemical purity and includes recommended storage conditions for powder and solutions.
  • High isotopic purity: 99.5%
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS
  • Solid, white to off-white appearance for easy handling
  • Defined storage stability for powder and in solvent to preserve integrity
  • Provides known molecular weight and formula for accurate quantitation
Catalog No. 50-002-83308 Supplier Medchemexpress LLC Supplier No. HYW131352S5MG
N-hexacosane-d54 is a deuterium-labeled analog of n-hexacosane designed for research applications as a tracer and internal standard in quantitative analyses such as NMR, GC-MS, and LC-MS. It is supplied as a solid with high isotopic and chemical purity and includes recommended storage conditions for powder and solutions.
  • High isotopic purity: 99.5%
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS
  • Solid, white to off-white appearance for easy handling
  • Defined storage stability for powder and in solvent to preserve integrity
  • Provides known molecular weight and formula for accurate quantitation