Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexadecyl sulfate, sodium salt +98%, Thermo Scientific™

Catalog No. AC385000250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC385000250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC385000250 Supplier Thermo Scientific Chemicals Supplier No. 385000250

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-01-0
Molecular Formula C16H33NaO3S
Molecular Weight (g/mol) 328.49
MDL Number MFCD00047766
InChI Key PNGBYKXZVCIZRN-UHFFFAOYSA-M
Synonym sodium hexadecyl sulfate, sodium cetyl sulfate, sodium n-hexadecyl sulfate, unii-3v3y3o7biq, conco sulfate c, avitex c, avitex sf, sodium hexyldecyl sulfate, tergitol anionic 7, 3v3y3o7biq
PubChem CID 23695542
IUPAC Name sodium hexadecane-1-sulfonate
SMILES [Na+].CCCCCCCCCCCCCCCCS([O-])(=O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3(CH2)15OSO3Na
Quantity 25 g
Formula Weight 344.49
Percent Purity ≥98.0% (from ash)
Physical Form Powder
Chemical Name or Material n-Hexadecyl sulfate, sodium salt
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.