Learn More
Medchemexpress LLC N'-Hexadecylthiophene-2-carbohydrazide | 2374313-54-7 | 99.43% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12891810MG
Encompass_Preferred
SIS17 is a mammalian histone deacetylase 11 (HDAC 11) inhibitor with an IC50 value of 0.83 μM. It inhibits the demyristoylation of serine hydroxymethyltransferase 2, a substrate of HDAC 11, but does not inhibit other HDACs. This product is for research use only.
  • IC50 value of 0.83 μM for HDAC11
  • Inhibits demyristoylation of serine hydroxymethyltransferase 2
  • Does not inhibit other HDACs
  • Increases fatty acylation level of SHMT2 in MCF7 cells
  • Demonstrates synergistic cytotoxicity with Oxaliplatin in K562 cells
  • White to off-white solid appearance
  • Purity of 99.43%
Catalog No. 50-003-41209