Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane for gas chromatography ECD and FID SupraSolv™, MilliporeSigma™

Catalog No. M1043714000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1043714000 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1043714000 Supplier MilliporeSigma Supplier No. 1.04371.4000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point −94.3°C
Density 0.66 g/cm3 (at 20°C)
Quantity 4 L
Explosion Limit 1.0 to 8.1% (V)
Flash Point −22°C
Ignition Point 240°C
Packaging Glass Bottle
Vapor Pressure 160 hPa (20°C)
Viscosity 0.50 mm2/s (20°C)
Boiling Point 69°C (1013 hPa)
Solubility Information 0.0095 g/L
Show More Show Less
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.