Learn More
Hach Company Hexane, for Organic Residue Analysis
Supplier:  Hach Company 2510253
Encompass
Low residue on evaporation makes this Hexane ideal for Oil & Grease analysis using EPA Method 1664A, Standard Methods 5520G and Hach Method 10300. 1-L bottle.
Catalog No. 50-199-6116